Learn More
Aviva Systems Biology Corp VPS26A Antibody - C-terminal region (ARP61669_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61669_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS26A. It was validated on Western Blot. . Alternate names: FLJ12930, HB58, Hbeta58, PEP8A, VPS26ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-436