Learn More
Medchemexpress LLC HY-12473 5mg Medchemexpress, Vps34-IN-2 CAS:1523404-29-6 Purity:>98%
SDP
Catalog No. 502081049
Encompass_Preferred
Medchemexpress, HY-12473 5mg Vps34-IN-2 CAS:1523404-29-6 Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1049 Supplier Medchemexpress LLC Supplier No. HY124735MG
Medchemexpress, HY-12473 5mg Vps34-IN-2 CAS:1523404-29-6 Vps34-IN-2 is a novel, potent and selective inhibitor of Vps34 with IC50s of 2 and 82 nM on the Vps34 enzymatic assay and the GFP-FYVE cellular assay, respectively. Vps34-IN-2 shows antiviral activity against SARS-CoV-2 (IC50 of 3.1 μM), HCoV-229E (IC50 of 0.7 μM) and HCoV-OC43. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.