Learn More
TARGETMOL CHEMICALS INC VPS34 inhibitor 1 Compound 19
SDP
Catalog No. 502877355
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. VPS34 inhibitor 1 (Compound 19, PIK-III analogue) (PIK-III analogue) is a potent and selective inhibitor of VPS34( IC50 15 nM) Purity 98.6%
Catalog No. 50-287-7355 Supplier TARGETMOL CHEMICALS INC Supplier No. T79445MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. VPS34 inhibitor 1 (Compound 19, PIK-III analogue) (PIK-III analogue) is a potent and selective inhibitor of VPS34( IC50 15 nM) Purity 98.6%