Learn More
STEMCELL Technologies Vps34-IN1, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 1000255
Encompass
Vps34-IN1 is a potent and reversible inhibitor (IC₅₀ = 25 nM) of vacuolar protein sorting 34 (Vps34), a class II phosphoinositide 3-kinase that regulates autophagosome formation (Bago et al.). CANCER RESEARCH· Reduces accumulation of phosphorylated phosphatidylinositol (PtdIns(3)P) at endosomal membranes (Bago et al.).
Catalog No. 50-204-5500