Learn More
Medchemexpress LLC Vps34-PIK-III | 1383716-40-2 | 99.6% | 2 MG
SDP
Catalog No. 5000340160
Encompass_Preferred
Vps34-PIK-III is an orally active and selective VPS34 inhibitor, demonstrating potent inhibition with an IC50 of 18 nM. It effectively inhibits autophagy and acts as a PI3K inhibitor, suppressing gene expression in liver cancer stem cells (CSCs). This compound serves as a valuable molecular tool for research.
  • Orally active and selective VPS34 inhibitor
  • Inhibits autophagy
  • Acts as a PI3K inhibitor
  • Suppresses gene expression in liver cancer stem cells
  • For research use only
Catalog No. 50-003-40160 Supplier Medchemexpress LLC Supplier No. HY127942MG
Vps34-PIK-III is an orally active and selective VPS34 inhibitor, demonstrating potent inhibition with an IC50 of 18 nM. It effectively inhibits autophagy and acts as a PI3K inhibitor, suppressing gene expression in liver cancer stem cells (CSCs). This compound serves as a valuable molecular tool for research.
  • Orally active and selective VPS34 inhibitor
  • Inhibits autophagy
  • Acts as a PI3K inhibitor
  • Suppresses gene expression in liver cancer stem cells
  • For research use only