Learn More
TARGETMOL CHEMICALS INC VPS34-PIK-III 5MG
SDP
Catalog No. 502576408
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vps34-PIK-III (VPS34-IN2) a selective inhibitor of VPS34 enzymatic activity inhibits autophagy and results in the stabilization of autophagy substrates. purity: 98%
Catalog No. 50-257-6408 Supplier TARGETMOL CHEMICALS INC Supplier No. T69455MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vps34-PIK-III (VPS34-IN2) a selective inhibitor of VPS34 enzymatic activity inhibits autophagy and results in the stabilization of autophagy substrates. purity: 98%