Learn More
Aviva Systems Biology Corp VPS37C Antibody - N-terminal region (ARP57073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS37C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VPS37C. Alternate names: FLJ20847ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62072