Learn More
Aviva Systems Biology Corp Vps45 Antibody - C-terminal region (ARP52315_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52315_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Vps45. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI462172, AW554165, mVps45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-703