Learn More
Biomatik Corporation Vps4A Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10753
Encompass_Preferred
Vps4A Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VPS4A. The Vps4A Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Vacuolar Protein Sorting-Associated Protein 4A Protein (1-120Aa). Applications: ELISA, WB, IHC. Target Synonyms: VPS4A. Accession Number: Q9UN37. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9259