Learn More
Aviva Systems Biology Corp VPS52 Antibody - middle region (ARP57644_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57644_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS52. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VPS52. Alternate names: ARE1, DKFZp547I194, RP5-1033B10, SAC2, SACM2L, dJ1033B10.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62919