Learn More
Aviva Systems Biology Corp VPS54 Antibody - N-terminal region (ARP56168_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56168_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS54. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VPS54. Alternate names: HCC8, SLP-8p, VPS54L, hVps54LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61744