Learn More
Aviva Systems Biology Corp VPS72 Antibody - N-terminal region (ARP33348_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33348_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS72. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VPS72. Alternate names: Swc2, TCFL1, YL-1, YL1, CFL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54974