Learn More
Aviva Systems Biology Corp VPS8 Antibody - N-terminal region (ARP42974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VPS8. Alternate names: FLJ32099, KIAA0804ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58381