Learn More
Germaine™ Laboratories VQPet Glucose Test Strips
Catalog No. 23111391
Change view
Click to view available options
Quantity:
50 Test Strips
Catalog No. Quantity
23-111-391 50 Test Strips
1 options
Catalog No. 23-111-391 Supplier Germaine™ Laboratories Supplier No. VGS01421
Add to Cart
Add to Cart
Intended as an aid for persons with diabetic pets and by veterinarians to monitor glucose concentration of cats and dogs and monitor the effectiveness of diabetes control.
  • Results in 5 Seconds
  • Only 0.5 μL Blood Sample Required from Pet = Less Pain for Pets
  • One Meter for Testing Both Cats & Dogs with Auto-calibration - No coding needed
  • Supports Alternative Site Testing for Pets
Specifications
Content And Storage 2°C to 30°C
Description Quantitatively measures the glucose concentration in venous blood and fresh capillary whole blood in pets
Detectable Analytes Glucose
Type Pet Glucose Monitoring
Includes 50 Test Strips
Quantity 50 Test Strips
Sample Size Only 0.5 μL Blood Sample Required from Pet = Less Pain for Pets
Test Time 5 sec.