Learn More
Medchemexpress LLC VQW-765 | 669770-29-0 | 99.6% | 294.39 | 25 MG
SDP
Catalog No. 5000403070
Encompass_Preferred
VQW-765 (AQW-051) is a selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist. It demonstrates anxiolytic-like effects in vivo and is suitable for research into anxiety disorders and acute performance anxiety.
  • Selective and orally active α7-nAChR agonist.
  • Demonstrates anxiolytic-like effects in vivo.
  • Suitable for research on anxiety disorder and acute performance anxiety.
  • Increases cognitive effect and learning/memory performance.
Catalog No. 50-004-03070 Supplier Medchemexpress LLC Supplier No. 25MGHY19490
VQW-765 (AQW-051) is a selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist. It demonstrates anxiolytic-like effects in vivo and is suitable for research into anxiety disorders and acute performance anxiety.
  • Selective and orally active α7-nAChR agonist.
  • Demonstrates anxiolytic-like effects in vivo.
  • Suitable for research on anxiety disorder and acute performance anxiety.
  • Increases cognitive effect and learning/memory performance.