Learn More
Medchemexpress LLC VQW-765 | 669770-29-0 | MFCD28502153 | 99.6% | 294.39 g/mol | C19H22N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1949010MG
Encompass_Preferred
VQW-765 is an orally active, selective partial agonist of the α7 nicotinic acetylcholine receptor (α7-nAChR) developed for preclinical research into anxiety and related CNS disorders. Supplied as a pre-weighed 10 mg research reagent, it is intended for use in in vitro and in vivo studies and is not for human consumption.
  • Selective α7 nicotinic acetylcholine receptor partial agonist.
  • Orally active compound suitable for in vivo dosing.
  • High chemical purity (approximately 99.6%).
  • Supplied as a pre-weighed 10 mg quantity for dosing convenience.
  • Appropriate for both in vitro and in vivo pharmacology studies.
  • Research use only; not for human or clinical use.
Catalog No. 50-002-94622