Learn More
Medchemexpress LLC VQW-765 (AQW-051) | 669770-29-0 | 99.58% | 294.39 | 5 MG
SDP
Catalog No. 5000401925
Encompass_Preferred
VQW-765 (AQW-051) is a selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist. It demonstrates a pKD value of 7.56 to recombinantly expressed human α7-nAChR and exhibits anxiolytic-like effects in vivo. This compound can be used for research into anxiety disorder and acute performance anxiety.
  • Selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist
  • Demonstrates potent agonist activity to calcium transients
  • Enhances cognitive effect and learning/memory performance
  • Shows anxiolytic-like effects in vivo
Catalog No. 50-004-01925 Supplier Medchemexpress LLC Supplier No. HY194905MG
VQW-765 (AQW-051) is a selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist. It demonstrates a pKD value of 7.56 to recombinantly expressed human α7-nAChR and exhibits anxiolytic-like effects in vivo. This compound can be used for research into anxiety disorder and acute performance anxiety.
  • Selective and orally active alpha-7 nicotinic acetylcholine receptor (α7-nAChR) agonist
  • Demonstrates potent agonist activity to calcium transients
  • Enhances cognitive effect and learning/memory performance
  • Shows anxiolytic-like effects in vivo