Learn More
NEOGEN Violet Red Bile Glucose Agar (ISO), 5 kg
SDP
Catalog No. 502015271
Encompass_Preferred
Violet Red Bile Glucose Agar is used for the enumeration of Enterobacteriaceae in foods and is not intended for use in the diagnosis of disease or other conditions in humans.Formulated according to ISO 21528:2017 (all parts), VRBGA is a modification of Violet Red Bile Lactose Agar (NCM0089) introduced by Mossel in 1978. VRBA contains lactose which is fermented by members of the coli/ aerogenes group, this medium gives a ‘coliform’ count. The VRBGA formula modification replaces lactose with glucose. Glucose is fermented by all members of the Enterobacteriaceae thus gives a presumptive Enterobacteriaceae count. Bile salts and crystal violet are used to inhibit Gram positive and non-enteric organisms. The overlay procedure ensures anaerobic conditions and suppresses the growth of non-fermentative Gram negative bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/violet-red-bile-glucose-agar-iso
Catalog No. 50-201-5271 Supplier NEOGEN Supplier No. 700004428
Violet Red Bile Glucose Agar is used for the enumeration of Enterobacteriaceae in foods and is not intended for use in the diagnosis of disease or other conditions in humans.Formulated according to ISO 21528:2017 (all parts), VRBGA is a modification of Violet Red Bile Lactose Agar (NCM0089) introduced by Mossel in 1978. VRBA contains lactose which is fermented by members of the coli/ aerogenes group, this medium gives a ‘coliform’ count. The VRBGA formula modification replaces lactose with glucose. Glucose is fermented by all members of the Enterobacteriaceae thus gives a presumptive Enterobacteriaceae count. Bile salts and crystal violet are used to inhibit Gram positive and non-enteric organisms. The overlay procedure ensures anaerobic conditions and suppresses the growth of non-fermentative Gram negative bacteria.For more information visit https://www.neogen.com/solutions/Microbiology/violet-red-bile-glucose-agar-iso