Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PolyCo™ VR™ Blue Gowns with Elastic Cuffs

Catalog No. 19125571
Encompass_Preferred
Change view
Click to view available options
Length (English):
50 in.
55 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-125-571 55 in.
19-125-570 50 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-125-571 Supplier PolyCo™ Supplier No. 42650
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gown material is 4 mil thick and provides protection for applications where the threat of biological or chemical contamination exist.

  • Durable 4 mil thickness
  • Lightweight
  • Reusable
  • Sleeves with elastic cuffs
  • Recyclable
  • Made in USA

Specifications

Length (English) 55 in.
Length (Metric) 127 cm
Packaging Type Bulk
Product Type Controlled Environments Gown
For Use With (Application) Food Processing, Critical and Controlled Environments, Healthcare, Manufacturing
Closure Type Ties
Sterility Non-sterile
Seam Type Welded
Sleeve Style Long Sleeve
Certifications/Compliance OSHA 29, CFR Part 1910.1030
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.