Learn More
NEOGEN Violet Red Bile Glucose Agar (VRBGA), 500 g
SDP
Catalog No. 502015216
Encompass_Preferred
Violet Red Bile Glucose Agar is used for the enumeration of Enterobacteriaceae in foods. Violet Red Bile Glucose Agar conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized USP/EP/JP for isolation and identification of bile-tolerant Gram-negative bacteria from non-sterile products. Conforms to Harmonized USP/EP/JP performance specification. Yeast extract provides vitamins and gelatin serves as source of carbon and nitrogen. Glucose is a fermentable carbohydrate and sodium chloride maintains the osmotic balance. Bile salts and crystal violet act as selective agents inhibiting many Gram-positive bacteria. The formulation is a modification of Violet Red Bile Agar by Mossel which substitutes lactose for glucose.For more information visit https://www.neogen.com/solutions/Microbiology/violet-red-bile-glucose-agar
Catalog No. 50-201-5216 Supplier NEOGEN Supplier No. 700003020
Violet Red Bile Glucose Agar is used for the enumeration of Enterobacteriaceae in foods. Violet Red Bile Glucose Agar conforms to Harmonized USP/EP/JP requirements and is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized USP/EP/JP for isolation and identification of bile-tolerant Gram-negative bacteria from non-sterile products. Conforms to Harmonized USP/EP/JP performance specification. Yeast extract provides vitamins and gelatin serves as source of carbon and nitrogen. Glucose is a fermentable carbohydrate and sodium chloride maintains the osmotic balance. Bile salts and crystal violet act as selective agents inhibiting many Gram-positive bacteria. The formulation is a modification of Violet Red Bile Agar by Mossel which substitutes lactose for glucose.For more information visit https://www.neogen.com/solutions/Microbiology/violet-red-bile-glucose-agar