Learn More
LICORbio VRDye™ 549 Donkey anti-Mouse IgG Secondary Antibody (0.5 mg)
Catalog No. NC1992336
Encompass_Preferred
Immunogen: Mouse lgG | Applications: VRDye™ 549 Donkey anti-Mouse IgG Secondary Antibody can be used for Western Blotting | Formulation: VRDye 549 secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C before and after reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
Catalog No. NC1992336 Supplier LICORbio Supplier No. 92654012
Immunogen: Mouse lgG | Applications: VRDye™ 549 Donkey anti-Mouse IgG Secondary Antibody can be used for Western Blotting | Formulation: VRDye 549 secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C before and after reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.