Learn More
LICORbio VRDye™ 549 Goat anti-Mouse IgG Secondary Antibody (0.5mg)
Catalog No. NC2209184
Encompass_Preferred
Immunogen: Mouse IgG paraproteins | Applications: Microscopy, Immunohistochemistry, and Flow Cytometry | Formulation: VRDye 549 secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.
This item is not returnable. View return policy
Catalog No. NC2209184 Supplier LICORbio Supplier No. 92654010
Immunogen: Mouse IgG paraproteins | Applications: Microscopy, Immunohistochemistry, and Flow Cytometry | Formulation: VRDye 549 secondary antibodies are supplied as purified immunoglobulin conjugates, lyophilized in phosphate-buffered saline, pH 7.4. Protect from light. Store at 4 °C prior to reconstitution. Each vial contains 10 mg/mL BSA (free of IgG and protease) as a stabilizer and 0.01% sodium azide as a preservative, after reconstitution. Concentration is 1.0 mg/mL when reconstituted as directed. Refer to the pack insert for details on reconstitution.