Learn More
Velp™ Scientific Vreceiver™ TKN Formula for UDK129 Distillation Unit
SDP
Catalog No. 11392149
Change view
Click to view available options
Capacity (Metric):
1 L
Catalog No. Capacity (Metric)
11-392-149 1 L
1 options
This item is not returnable. View return policy
Catalog No. 11-392-149 Supplier Velp™ Scientific Supplier No. A00000411
Add to Cart
Add to Cart
This item is not returnable. View return policy
Velp™'s unique formula for use with the UDK129 Distillation Unit.
  • Certified formula composed of Boric Acid powder and a mixture of indicators mentioned by AOAC methods (Bromocresol green and Methyl red)
  • Allows for fast and standardized receiving solution preparation for colorimetric titration
Specifications
Type Powder
Capacity (Metric) 1 L
For Use With (Equipment) UDK129 Distillation Unit
Quantity 10 Pk.