Learn More
Medchemexpress LLC HY-126542 5mg , VRK-IN-1 CAS:2378855-09-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1265425MG
Encompass_Preferred
HY-126542 5mg VRK-IN-1 CAS: 2378855-09-3 VRK-IN-1 is a potent and selective inhibitor of vaccinia-related kinases 1 (VRK1), with an IC50 of 150 nM. VRK1 is human Ser/Thr protein kinases associated with increased cell division and neurological disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9133