Learn More
Carnabio Usa Inc VRK1/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0311010
Encompass
VRK1, Full-length, Wild type, Amino Acid 1-396, NP_003375.1, Expressed in E. coli; 10microg
Catalog No. 50-754-1023