Learn More
Carnabio Usa Inc VRK1/5UG/FULL-LENGTH
Catalog No. 507540657
Encompass
VRK1, Full-length, Wild type, Amino Acid 1-396, NP_003375.1, Expressed in E. coli; 5microg
Catalog No. 50-754-0657 Supplier Carnabio Usa Inc Supplier No. 031105
VRK1, Full-length, Wild type, Amino Acid 1-396, NP_003375.1, Expressed in E. coli; 5microg