Learn More
Eurofins Discoverx VRK2, active, 10 µg. (14-732)
Supplier:  Eurofins Discoverx 14732
Encompass
N-terminal 6xHis-tagged recombinant, human VRK2 amino acids 1–336. VRK2 is a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This protein is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, leukocytes, fetal liver, and carcinomas. Its protein localizes to the endoplasmic reticulum and has been shown to phosphorylate casein and undergo auto phosphorylation. Multiple alternatively spliced transcript variants have been found for this gene. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9253