Learn More
Medchemexpress LLC VRT-043198 | 244133-31-1 | 98.89% | 480.94 | 50 MG
SDP
Catalog No. 5000437983
Encompass_Preferred
VRT-043198 is a potent, selective, and blood-brain barrier permeable inhibitor of interleukin-converting enzyme/caspase-1 subfamily caspases. It is the agent metabolite of VX-765 (Belnacasan). This inhibitor exhibits Ki values of 0.8 nM for ICE/caspase-1 and 0.6 nM for caspase-4.
  • Potent, selective, and blood-brain barrier permeable inhibitor of interleukin-converting enzyme/caspase-1 subfamily caspases.
  • Exhibits Ki values of 0.8 nM for ICE/caspase-1 and 0.6 nM for caspase-4.
  • Exhibits 100- to 10,000-fold selectivity against other caspase-3 and -6 to -9.
  • Inhibits the release of interleukin (IL)-1β and IL-18.
  • Has little effect on IL-1α, tumor necrosis factor-, IL-6, and IL-8.
  • Lacks potent antiapoptotic activity.
Catalog No. 50-004-37983 Supplier Medchemexpress LLC Supplier No. HY11222650MG
VRT-043198 is a potent, selective, and blood-brain barrier permeable inhibitor of interleukin-converting enzyme/caspase-1 subfamily caspases. It is the agent metabolite of VX-765 (Belnacasan). This inhibitor exhibits Ki values of 0.8 nM for ICE/caspase-1 and 0.6 nM for caspase-4.
  • Potent, selective, and blood-brain barrier permeable inhibitor of interleukin-converting enzyme/caspase-1 subfamily caspases.
  • Exhibits Ki values of 0.8 nM for ICE/caspase-1 and 0.6 nM for caspase-4.
  • Exhibits 100- to 10,000-fold selectivity against other caspase-3 and -6 to -9.
  • Inhibits the release of interleukin (IL)-1β and IL-18.
  • Has little effect on IL-1α, tumor necrosis factor-, IL-6, and IL-8.
  • Lacks potent antiapoptotic activity.