Learn More
Medchemexpress LLC HY-112226 5mg , VRT-043198 CAS:244133-31-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1122265MG
Encompass_Preferred
HY-112226 5mg VRT-043198 CAS: 244133-31-1 VRT-043198, the drug metabolite of VX-765 (Belnacasan), is a potent, selective and blood-brain barrier permeable inhibitor of interleukin-converting enzyme/caspase-1 subfamily caspases. VRT-043198 exhibits Ki values of 0.8 nM and 0.6 nM for ICE/caspase-1 and caspase-4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0548