Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VRT-532 10mM 1mL | 38214-71-0 | 1 ML
SDP

Catalog No. 5000228993
Encompass_Preferred

VRT-532 is a small-molecule CFTR modulator supplied as a ready-to-use 10 mM solution in DMSO. It enhances channel activity and intrinsic ATPase activity of G551D-CFTR and is used in cystic fibrosis research and related cellular assays.

  • Potent CFTR modulator that enhances G551D-CFTR channel and ATPase activity.
  • Supplied as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity appropriate for research use (reported 98.98%).
  • Chemical formula C16H14N2O; molecular weight ~250.3 g/mol.
  • Storage: powder at -20°C for up to 3 years; solutions at -80°C for up to 6 months.

Catalog No. 50-002-28993 Supplier Medchemexpress LLC Supplier No. HY11472510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VRT-532 is a small-molecule CFTR modulator supplied as a ready-to-use 10 mM solution in DMSO. It enhances channel activity and intrinsic ATPase activity of G551D-CFTR and is used in cystic fibrosis research and related cellular assays.

  • Potent CFTR modulator that enhances G551D-CFTR channel and ATPase activity.
  • Supplied as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity appropriate for research use (reported 98.98%).
  • Chemical formula C16H14N2O; molecular weight ~250.3 g/mol.
  • Storage: powder at -20°C for up to 3 years; solutions at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.