Learn More
TARGETMOL CHEMICALS INC VRT-532 5MG
SDP
Catalog No. 502580461
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VRT-532 (CFpot-532) is an effective modulator of CFTR and is commonly used in studies of cystic fibrosis (CF) caused by CFTR defects. Arylsulfatase B (ARSB) which is necessary to reduce the accumulation of sulphate glycosaminosaccharide (gag) in cystic fibrosis and modification of CFTR by small molecule modulator VRT-532 can increase ARSB and reduce the accumulation of 4-chondroitin sulfate. purity: 99%
Catalog No. 50-258-0461 Supplier TARGETMOL CHEMICALS INC Supplier No. T291145MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VRT-532 (CFpot-532) is an effective modulator of CFTR and is commonly used in studies of cystic fibrosis (CF) caused by CFTR defects. Arylsulfatase B (ARSB) which is necessary to reduce the accumulation of sulphate glycosaminosaccharide (gag) in cystic fibrosis and modification of CFTR by small molecule modulator VRT-532 can increase ARSB and reduce the accumulation of 4-chondroitin sulfate. purity: 99%