Learn More
Medchemexpress LLC VRT-534 | 926664-32-6 | 98.3% | 497.99 | 25 MG
SDP
Catalog No. 5000363988
Encompass_Preferred
VRT-534 is a chemical chaperone that specifically targets connexin 26 (Cx26). It demonstrates dose-responsive binding to recombinant wild-type Cx26 and mutant Cx26 K188N. This compound can be utilized in research pertaining to hearing impairment and has shown restorative effects on Cx26 mutants in HeLa cells.
  • Exhibits dose-responsive binding to Cx26
  • Restores vitality of HeLa cells
  • Useful in research for hearing impairment
Catalog No. 50-003-63988 Supplier Medchemexpress LLC Supplier No. 25MGHY149318
VRT-534 is a chemical chaperone that specifically targets connexin 26 (Cx26). It demonstrates dose-responsive binding to recombinant wild-type Cx26 and mutant Cx26 K188N. This compound can be utilized in research pertaining to hearing impairment and has shown restorative effects on Cx26 mutants in HeLa cells.
  • Exhibits dose-responsive binding to Cx26
  • Restores vitality of HeLa cells
  • Useful in research for hearing impairment