Learn More
Medchemexpress LLC VRT-534 | 926664-32-6 | 98.32% | 497.99 | 100 MG
SDP
Catalog No. 5000361189
Encompass_Preferred
VRT-534 (Compound 3) is a chemical chaperone that targets connexin 26 (Cx26). It demonstrates dose-responsive binding to recombinant WT Cx26 and mutant Cx26 K188N with EC50s of 19 and 5 μM, respectively. This compound can be utilized in research related to hearing impairment.
  • Chemical chaperone targeting connexin 26 (Cx26).
  • Exhibits dose-responsive binding to recombinant WT Cx26 and mutant Cx26 K188N.
  • Can be used in research related to hearing impairment.
  • Restores the vitality of HeLa cells.
Catalog No. 50-003-61189 Supplier Medchemexpress LLC Supplier No. HY149318100MG
VRT-534 (Compound 3) is a chemical chaperone that targets connexin 26 (Cx26). It demonstrates dose-responsive binding to recombinant WT Cx26 and mutant Cx26 K188N with EC50s of 19 and 5 μM, respectively. This compound can be utilized in research related to hearing impairment.
  • Chemical chaperone targeting connexin 26 (Cx26).
  • Exhibits dose-responsive binding to recombinant WT Cx26 and mutant Cx26 K188N.
  • Can be used in research related to hearing impairment.
  • Restores the vitality of HeLa cells.