Learn More
Cayman Chemical VRy LNgchaIn Saturated Fat 1mL
SDP
Catalog No. 502988130
Encompass_Preferred
A mixture of five very long-chain saturated fatty acid methyl esters; intended for use as a reference standard for the quantification of very long-chain saturated fatty acids in various sample types by GC-MS
Catalog No. 50-298-8130 Supplier Cayman Chemical Supplier No. 293701 ML
A mixture of five very long-chain saturated fatty acid methyl esters; intended for use as a reference standard for the quantification of very long-chain saturated fatty acids in various sample types by GC-MS