Learn More
Beckman Coulter Beckman Coulter BioSafe Round Bucket Lids, Set of 2 - For VS 4.750 Swinging-Bucket Rotor (Allegra V-15R, 4 x 750 mL Buckets)
Catalog No. NC2583592 Shop All Beckman Coulter Products
Encompass
Beckman Coulter BioSafe round bucket lids (set of 2) provide containment for 750 mL round buckets used with VS 4.750 swinging-bucket rotors on Allegra V-15R centrifuges. They are included in BioSafe rotor packages and support biocertification when used as specified by the manufacturer.
  • Provides BioSafe coverage for 750 mL round buckets.
  • Sold as a set of two lids.
  • Compatible with 4 x 750 mL swinging-bucket rotors and round buckets.
  • Included in blood sample and BioSafe swinging bucket packages.
  • Helps reduce aerosol and spill contamination during centrifugation.
Catalog No. NC2583592 Supplier Beckman Coulter Supplier No. C63153
Beckman Coulter BioSafe round bucket lids (set of 2) provide containment for 750 mL round buckets used with VS 4.750 swinging-bucket rotors on Allegra V-15R centrifuges. They are included in BioSafe rotor packages and support biocertification when used as specified by the manufacturer.
  • Provides BioSafe coverage for 750 mL round buckets.
  • Sold as a set of two lids.
  • Compatible with 4 x 750 mL swinging-bucket rotors and round buckets.
  • Included in blood sample and BioSafe swinging bucket packages.
  • Helps reduce aerosol and spill contamination during centrifugation.