Learn More
Selleck Chemical LLC VS-5584
SDP
Catalog No. 5000002043
Encompass_Preferred
VS-5584 (SB2343) is a potent and selective dual PI3K/mTOR inhibitor for mTOR PI3K / / / with IC50 of 3 4 nM and 2 6-21 nM respectively Phase 1
Catalog No. 50-000-02043 Supplier Selleck Chemical LLC Supplier No. S701610MG
VS-5584 (SB2343) is a potent and selective dual PI3K/mTOR inhibitor for mTOR PI3K / / / with IC50 of 3 4 nM and 2 6-21 nM respectively Phase 1