Learn More
Cayman Chemical VS-5584 nalog 10mg
SDP
Catalog No. 502991285
Encompass_Preferred
A dual inhibitor of PI3Kα and mTOR (IC50s = 17 and 150 nM, respectively); a desmethyl analog of VS-5584
Catalog No. 50-299-1285 Supplier Cayman Chemical Supplier No. 3032410 MG
A dual inhibitor of PI3Kα and mTOR (IC50s = 17 and 150 nM, respectively); a desmethyl analog of VS-5584