Learn More
Cayman Chemical VS-5584 analog, Purity: ≥98%, CAS: 1246535-95-4, 25 mg
SDP
Catalog No. 502991292
Encompass_Preferred
A dual inhibitor of PI3Kα and mTOR (IC50s = 17 and 150 nM, respectively); a desmethyl analog of VS-5584
Catalog No. 50-299-1292 Supplier Cayman Chemical Supplier No. 3032425 MG
A dual inhibitor of PI3Kα and mTOR (IC50s = 17 and 150 nM, respectively); a desmethyl analog of VS-5584