Learn More
Apexbio Technology LLC VS-5584 (SB2343) 1246560-33-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064031
Encompass_Preferred
VS-5584 (SB2343 CAS 1246560-33-7) is a purine-based inhibitor targeting the mammalian target of rapamycin (mTOR) and all class I phosphoinositide 3-kinase (PI3K) isoforms (PI3K PI3K PI3K and PI3K ) It functions through ATP-competitive inhibition displaying IC50 values of 37 16 68 25 and 42 nmol/L against mTOR PI3K PI3K PI3K and PI3K respectively Through inhibition of PI3K and mTORC1/2-mediated substrate phosphorylation VS-5584 modulates the PI3K/mTOR signaling pathway frequently dysregulated in cancers In preclinical tumor xenograft studies VS-5584 has demonstrated dose-dependent inhibition of tumor growth and pathway signaling underscoring its utility in oncology research
Catalog No. 50-000-64031 Supplier Apexbio Technology LLC Supplier No. A39275.1
 Restricted Item
Add to Cart
Add to Cart
VS-5584 (SB2343 CAS 1246560-33-7) is a purine-based inhibitor targeting the mammalian target of rapamycin (mTOR) and all class I phosphoinositide 3-kinase (PI3K) isoforms (PI3K PI3K PI3K and PI3K ) It functions through ATP-competitive inhibition displaying IC50 values of 37 16 68 25 and 42 nmol/L against mTOR PI3K PI3K PI3K and PI3K respectively Through inhibition of PI3K and mTORC1/2-mediated substrate phosphorylation VS-5584 modulates the PI3K/mTOR signaling pathway frequently dysregulated in cancers In preclinical tumor xenograft studies VS-5584 has demonstrated dose-dependent inhibition of tumor growth and pathway signaling underscoring its utility in oncology research