Learn More
Beckman Coulter TCR VS2-FITC, IMMU 389, 1 mL, ASR
Catalog No. NC1908649
Encompass_Preferred
Vδ2 is the human variable δ2 gene of the γδ T cell receptor. Vδ2+ T cells are the main population of the circulating γδ T cells. It is expressed by 0.5% to 10% of PBL in normal blood, representing around 70% of the peripheral γδ T cells. Vδ2 is mostly associated with Vγ9. This sequence is also referred to as TRDV2 (based on the IMGT gene nomenclature).
Catalog No. NC1908649 Supplier Beckman Coulter Supplier No. IM1464
Vδ2 is the human variable δ2 gene of the γδ T cell receptor. Vδ2+ T cells are the main population of the circulating γδ T cells. It is expressed by 0.5% to 10% of PBL in normal blood, representing around 70% of the peripheral γδ T cells. Vδ2 is mostly associated with Vγ9. This sequence is also referred to as TRDV2 (based on the IMGT gene nomenclature).