Learn More
Aviva Systems Biology Corp VSIG1 Antibody - N-terminal region (ARP55832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VSIG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VSIG1. Alternate names: 1700062D20Rik, GPA34, MGC44287, dJ889N15.1ApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-61628