Learn More
ACROBiosystems Recombinant Protein;Human VSIG2 Protein;HEK293;1MG;VS2,VSIG2,CT-like protein,CTH,CTXL,VSIG-2
Catalog No. 501658334
Encompass_Preferred
Recombinant Protein;Human VSIG2 Protein;1MG;Human VSIG2, His Tag, Strep Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Ala 243 (Accession # AAH07313).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8334 Supplier ACROBiosystems Supplier No. VS2H52251MG
Recombinant Protein;Human VSIG2 Protein;1MG;Human VSIG2, His Tag, Strep Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Ala 243 (Accession # AAH07313).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.