Learn More
ACROBiosystems ACROBiosystems Human VSIG3 Protein, Fc Tag
Supplier:  ACROBiosystems VS3H52581MG
Encompass_Preferred
Recombinant Protein;1MG;Human VSIG3, Fc Tag (VS3-H5258) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accession # Q5DX21-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.7 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VSIG3, Fc Tag (Cat. No. VS3-H5258) at 5 μg/mL (100 μL/well) can bind Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B75-H5258) with a linear range of 0.313-10 μg/mL (Routinely tested).;Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B75-H5258) Captured on CM5 chip via anti-mouse antibodies surface can bind Human VSIG3, Fc Tag (Cat. No. VS3-H5258) with an affinity constant of 8.87 μM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-6834