Learn More
Sino Biological Human, VSIG4 cDNA ORF Clone, C-GFPSpark tag
Catalog No. NC1699031
Encompass_Preferred
Full length Clone DNA of Human V-set and immunoglobulin domain containing 4 with C terminal GFPSpark tag.
Catalog No. NC1699031 Supplier Sino Biological Supplier No. HG12163ACG
Full length Clone DNA of Human V-set and immunoglobulin domain containing 4 with C terminal GFPSpark tag.