Learn More
Medchemexpress LLC VSIG4 Human HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7142450UG
Encompass_Preferred
VSIG4 Protein, Human (HEK293, His) is a recombinant extracellular region of human VSIG4 expressed in HEK293 cells and supplied with a C-terminal His tag. The 264-amino-acid construct (R20-P283) is provided in convenient microgram quantities for biochemical and cell-based research, including binding and functional assays. Intended for research use only.
  • Recombinant extracellular region expressed in HEK293 cells.
  • C-terminal His tag for straightforward purification.
  • 264 amino acids (R20-P283) matching the extracellular construct.
  • Available in 20 μg, 50 μg, and 100 μg sizes.
  • Suitable for in vitro binding and functional assays.
  • For research use only, not for therapeutic applications.
Catalog No. 50-001-95653