Learn More
ACROBiosystems Recombinant Protein;Human VSIG4 Protein;HEK293;1MG;VS4,VSIG4,CRIg,Z39IG,VSIG-4
Catalog No. 501658336
Encompass_Preferred
Recombinant Protein;Human VSIG4 Protein;1MG;Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 283 (Accession # AAH10525).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8336 Supplier ACROBiosystems Supplier No. VS4H52261MG
Recombinant Protein;Human VSIG4 Protein;1MG;Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 283 (Accession # AAH10525).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30 kDa. The protein migrates as 45-48 kDa under reducing (R) condition (SDS-PAGE).