Learn More
ACROBiosystems Recombinant Protein;Human VSIG8 Protein, Fc tag;HEK293;1MG;VS8,VSIG8,C1orf204
Catalog No. 501658428
Encompass_Preferred
Recombinant Protein;Human VSIG8 Protein, Fc tag;1MG;Human VSIG8, Fc Tag (VS8-H5250) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.7 kDa. As a result of glycosylation, the protein migrates as 55-65 kDa under reducing (R) condition, and 110-130 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8428 Supplier ACROBiosystems Supplier No. VS8H52501MG
Recombinant Protein;Human VSIG8 Protein, Fc tag;1MG;Human VSIG8, Fc Tag (VS8-H5250) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53.7 kDa. As a result of glycosylation, the protein migrates as 55-65 kDa under reducing (R) condition, and 110-130 kDa under non-reducing (NR) condition (SDS-PAGE).