Learn More
ACROBiosystems Recombinant Protein;Human VSIG8 Protein;HEK293;1MG;VS8,VSIG8,C1orf204
Catalog No. 501658430
Encompass_Preferred
Recombinant Protein;Human VSIG8 Protein;1MG;Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29 kDa. The protein migrates as 35-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VSIG8, His Tag (Cat. No. VS8-H52H1) at 10 μg/mL (100 μL/well) can bind Human B7-H5, Fc Tag (Cat. No. B75-H5259) with a linear range of 0.625-20 μg/mL (QC tested).
Catalog No. 50-165-8430 Supplier ACROBiosystems Supplier No. VS8H52H11MG
Recombinant Protein;Human VSIG8 Protein;1MG;Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29 kDa. The protein migrates as 35-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VSIG8, His Tag (Cat. No. VS8-H52H1) at 10 μg/mL (100 μL/well) can bind Human B7-H5, Fc Tag (Cat. No. B75-H5259) with a linear range of 0.625-20 μg/mL (QC tested).