Learn More
Ace Glass, Inc. Vessel, ultrasonic, jacketed, 10-50mL, #25 center, (2)14/20 sides, body only
SDP
Catalog No. 50977142
Encompass
10-50mL jacketed ultrasonic reaction vessel body only. Vessel has a #25 center neck, and (2) 14/20 side necks. Jacket provides active cooling from tap water or a mechanical cooler, and is cylindrical with a flat bottom for greater stability. Vessel has hose connections for 3/8 ID tubing.
Catalog No. 50-977-142 Supplier Ace Glass, Inc. Supplier No. 985012
10-50mL jacketed ultrasonic reaction vessel body only. Vessel has a #25 center neck, and (2) 14/20 side necks. Jacket provides active cooling from tap water or a mechanical cooler, and is cylindrical with a flat bottom for greater stability. Vessel has hose connections for 3/8 ID tubing.