Learn More
Medchemexpress LLC VSLRGDTRG acetate | 99.4% | 960.05 | 5 MG
SDP
Catalog No. 5000423054
Encompass_Preferred
VSLRGDTRG acetate is a synthetic peptide containing the RGD motif from cadherin 17 (CDH17), which binds to α2β1 integrin and activates its signaling pathway. It promotes the high-affinity conformational change of β1 integrin through the RGD motif, enhancing cell adhesion and phosphorylation of FAK and ERK1/2, thereby driving tumor proliferation and metastasis. VSLRGDTRG acetate can be used in research on cancers expressing CDH17, such as colon cancer and pancreatic cancer.
  • Binds to α2β1 integrin and activates its signaling pathway.
  • Promotes the high-affinity conformational change of β1 integrin through the RGD motif.
  • Enhances cell adhesion and phosphorylation of FAK and ERK1/2.
  • Drives tumor proliferation and metastasis.
  • Can be used in research on cancers expressing CDH17, such as colon cancer and pancreatic cancer.
Catalog No. 50-004-23054 Supplier Medchemexpress LLC Supplier No. HYP10941A5MG
VSLRGDTRG acetate is a synthetic peptide containing the RGD motif from cadherin 17 (CDH17), which binds to α2β1 integrin and activates its signaling pathway. It promotes the high-affinity conformational change of β1 integrin through the RGD motif, enhancing cell adhesion and phosphorylation of FAK and ERK1/2, thereby driving tumor proliferation and metastasis. VSLRGDTRG acetate can be used in research on cancers expressing CDH17, such as colon cancer and pancreatic cancer.
  • Binds to α2β1 integrin and activates its signaling pathway.
  • Promotes the high-affinity conformational change of β1 integrin through the RGD motif.
  • Enhances cell adhesion and phosphorylation of FAK and ERK1/2.
  • Drives tumor proliferation and metastasis.
  • Can be used in research on cancers expressing CDH17, such as colon cancer and pancreatic cancer.